Merck
A simple method for quantitative determination of polysaccharides in fungal cell walls.
Nature protocols (2007-04-05)
Jean Marie François
ABSTRAKT
A simple and reliable method for quantitative determination of cell wall polymers in fungal cell with an s.e.m. of 5% is described. This protocol is based on the hydrolysis by sulfuric acid of beta-glucan, mannan, galactomannan and chitin present at different levels in the wall of yeasts and filamentous fungi into their corresponding monomers glucose, mannose, galactose and glucosamine. The released monosaccharides are subsequently separated and quantified by high-performance ionic chromatography coupled to pulse amperometry detection, with a detection limit of 1.0 mug ml(-1). This procedure is well suited to screening a large collection of yeast mutants or to evaluating effects of environmental conditions on cell wall polysaccharide content. This procedure is also applicable to other fungal species, including Schizosaccharomyces pombe, Candida albicans and Aspergillus fumigatus. Results can be obtained in 3 d.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chitin from shrimp shells, BioReagent, suitable for analysis of chitinase, purified powder