Merck
Using Tandem Affinity Purification to Identify Circadian Clock Protein Complexes from Arabidopsis.
Methods in molecular biology (Clifton, N.J.) (2021-10-22)
Maria L Sorkin, Dmitri A Nusinow
ABSTRAKT
Identification of protein-protein interactions is an effective method of elucidating new roles for circadian clock-associated proteins that can expand beyond the information collected from transcriptional studies and genetic screens. Tandem affinity purification coupled with liquid chromatography mass spectrometry (APMS) utilizes epitope-tagged versions of your protein of interest to co-precipitate direct and indirect protein partners. Here, we provide a protocol and suggestions for proper design of 6x-His-3x-FLAG-tagged clock proteins and isolation of protein-protein interactions using two immunoprecipitation steps for increased specificity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)