Merck
Cell cycle synchronization of tobacco BY-2 cells.
Nature protocols (2007-04-05)
Fumi Kumagai-Sano, Tomomi Hayashi, Toshio Sano, Seiichiro Hasezawa
ABSTRAKT
Synchronization is a powerful technique for understanding cell cycle events. Here, we describe the procedure for synchronizing tobacco bright yellow 2 (BY-2) cell line, with which an exceptionally high level of synchrony can be achieved. It basically relies on an "arrest-and-release" strategy using aphidicolin, an inhibitor of DNA replication, and propyzamide, a plant-microtubule disruptant. In a single-step process using aphidicolin alone, a cell population with about 70% of the cells at mitosis can be achieved, whereas by a two-step method using the two inhibitors sequentially, the level of synchrony can reach over 90%. The method of choice depends not only on the peak mitotic cell proportion but also on the cell cycle stage that is targeted for analysis. Both procedures take about 1.5 days, and cell cycle progression can be observed from the S phase to the next G1 phase at about 12 h after a 24 h-period treatment with aphidicolin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized