- Characterization of protein-protein interactions: application to the understanding of peroxisome biogenesis.
Characterization of protein-protein interactions: application to the understanding of peroxisome biogenesis.
Methods in molecular biology (Clifton, N.J.) (2007-10-24)
Sebastien Leon, Ivet Suriapranata, Mingda Yan, Naganand Rayapuram, Amar Patel, Suresh Subramani
PMID17951646
ABSTRAKT
With the approaching completion of the Pichia pastoris genome, a greater emphasis will have to be placed on the proteome and the protein-protein interactions between its constituents. This chapter discusses methods that have been used for the study of such interactions among both soluble and membrane-associated proteins in peroxisome biogenesis. The procedures are equally applicable to other cellular processes.