Merck
An improved explants culture method: Sustainable isolation of keloid fibroblasts with primary characteristics.
Journal of cosmetic dermatology (2022-09-29)
Yan He, Shengpeng Zhang, Wu Bao, Yi Xue, Bin Yin, Xialin Cheng, Zexin Zhang, Chiyu Jia
ABSTRAKT
Keloid (KD) is a complex fibroproliferative disease, but the exact mechanisms underlying keloid pathogenesis remain to be elucidated. The primary keloid fibroblasts (KFs) culture in vitro has always been a fundamental measure to study the pathogenesis of keloid. However, the traditional primary culture methods have some limitations, such as a long culture cycle, low specimen utilization rate and so on. Improve the keloid explants culture method sts. We proposed an improved new "explants multiple culture method"-reusing keloid explants for primary culture and harvesting the primary KFs in specific culture times. Meanwhile, the purity, proliferation, apoptosis, migration, invasion, extracellular matrix synthesis, and some fibrosis and inflammation-related proteins of KFs obtained from the first, fifth, and tenth explants cultures were detected. The results showed that the culture cycle of this new method (Cell Isolation: 2.67 ± 0.86 days, Explants removal: 8.83 ± 0.79 days, Cell Passage: 15.17 ± 1.39 days) was significantly shorter than that of the traditional method (Cell Isolation: 8.67 ± 1.84 days, Explants removal: 17.67 ± 2.17 days, Cell Passage: 22.67 ± 1.84 days). No significant difference was observed between the phenotypes of the fibroblasts obtained from the first explants culture and cultures less than 10 times (p > 0.05). Taken together, this study provides an effective method for the primary culture of KFs with a higher specimen utilization rate and shorter culture cycle. This method breaks through the limitation of traditional explants culture requiring a large number of keloid specimens and provides a rich source of KFs for the study of keloid.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture