Merck
m6A-SAC-seq for quantitative whole transcriptome m6A profiling.
Nature protocols (2022-11-27)
Ruiqi Ge, Chang Ye, Yong Peng, Qing Dai, Yutao Zhao, Shun Liu, Pingluan Wang, Lulu Hu, Chuan He
ABSTRAKT
N6-methyladenosine (m6A) is the most abundant mRNA modification in mammalian cells, regulating many physiological processes. Here we describe a method for base-resolution, quantitative m6A sequencing in the whole transcriptome. The enzyme and small-molecule cofactor used in this protocol are prepared by recombinant protein expression and organic synthesis, respectively. Then the library can be prepared from various types of RNA samples using a ligation-based strategy, with m6A modifications being labeled by the enzyme and cofactor. Detailed instructions on ensuing data analysis are also included in this protocol. The method generates highly reproducible results, uncovering 31,233-129,263 sites using as little as 2 ng of poly A+ RNA. These identified sites correspond well with previous m6A profiling results, covering over 65% of peaks detected by the antibody-based approaches. Compared with other currently available methods, this method can be applied to various types of biological samples, including fresh and frozen tissues as well as formalin-fixed paraffin-embedded samples, providing a quantitative method to uncover new insights into m6A biology. The protocol requires basic expertise in molecular biology, recombinant protein expression and organic synthesis. The whole protocol can be done in 15 days, with the library preparation taking 5 days.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
DNaza I, from bovine pancreas
Sigma-Aldrich
SUMO Protease, His tagged recombinant protein, lyophilized powder
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%