Merck
High cell density and high-resolution 3D bioprinting for fabricating vascularized tissues.
Science advances (2023-02-23)
Shangting You, Yi Xiang, Henry H Hwang, David B Berry, Wisarut Kiratitanaporn, Jiaao Guan, Emmie Yao, Min Tang, Zheng Zhong, Xinyue Ma, Daniel Wangpraseurt, Yazhi Sun, Ting-Yu Lu, Shaochen Chen
ABSTRAKT
Three-dimensional (3D) bioprinting techniques have emerged as the most popular methods to fabricate 3D-engineered tissues; however, there are challenges in simultaneously satisfying the requirements of high cell density (HCD), high cell viability, and fine fabrication resolution. In particular, bioprinting resolution of digital light processing-based 3D bioprinting suffers with increasing bioink cell density due to light scattering. We developed a novel approach to mitigate this scattering-induced deterioration of bioprinting resolution. The inclusion of iodixanol in the bioink enables a 10-fold reduction in light scattering and a substantial improvement in fabrication resolution for bioinks with an HCD. Fifty-micrometer fabrication resolution was achieved for a bioink with 0.1 billion per milliliter cell density. To showcase the potential application in tissue/organ 3D bioprinting, HCD thick tissues with fine vascular networks were fabricated. The tissues were viable in a perfusion culture system, with endothelialization and angiogenesis observed after 14 days of culture.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TissueFab® bioink DLP, synthetic, Vis/405nm
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Biotusz TissueFab DLP, with RGD, synthetic, Vis/405nm