Merck
EXOSC10/Rrp6 is essential for the eight-cell embryo/morula transition.
Developmental biology (2021-12-30)
Fabrice G Petit, Soazik P Jamin, Pierre-Yves Kernanec, Emmanuelle Becker, Guillaume Halet, Michael Primig
ABSTRAKT
The conserved 3'-5' exoribonuclease EXOSC10/Rrp6 is required for gametogenesis, brain development, erythropoiesis and blood cell enhancer function. The human ortholog is essential for mitosis in cultured cancer cells. Little is known, however, about the role of Exosc10 during embryo development and organogenesis. We generated an Exosc10 knockout model and find that Exosc10-/- mice show an embryonic lethal phenotype. We demonstrate that Exosc10 maternal wild type mRNA is present in mutant oocytes and that the gene is expressed during all stages of early embryogenesis. Furthermore, we observe that EXOSC10 early on localizes to the periphery of nucleolus precursor bodies in blastomeres, which is in keeping with the protein's role in rRNA processing and may indicate a function in the establishment of chromatin domains during initial stages of embryogenesis. Finally, we infer from genotyping data for embryonic days e7.5, e6.5 and e4.5 and embryos cultured in vitro that Exosc10-/- mutants arrest at the eight-cell embryo/morula transition. Our results demonstrate a novel essential role for Exosc10 during early embryogenesis, and they are consistent with earlier work showing that impaired ribosome biogenesis causes a developmental arrest at the morula stage.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid
Sigma-Aldrich
Monoclonal Anti-PSF antibody produced in mouse, clone B92, purified from hybridoma cell culture
Sigma-Aldrich
Anti-B23 antibody, Mouse monoclonal, clone FC82291, purified from hybridoma cell culture