- Generation of human iPSC-derived neurofibromaspheres for in vitro and in vivo uses.
Generation of human iPSC-derived neurofibromaspheres for in vitro and in vivo uses.
STAR protocols (2023-03-29)
Helena Mazuelas, Itziar Uriarte-Arrazola, Juana Fernández-Rodríguez, Miriam Magallón-Lorenz, Alberto Villanueva, Conxi Lázaro, Bernat Gel, Eduard Serra, Meritxell Carrió
PMID36976677
ABSTRAKT
Neurofibromas are benign peripheral nervous system tumors associated with neurofibromatosis type 1, which originate from NF1(-/-) Schwann cell precursors. We describe a protocol to generate neurofibromaspheres by differentiating NF1(-/-) Schwann cells from induced pluripotent stem cells and combining them with neurofibroma primary fibroblasts. We also describe the development of neurofibroma-like tumors when neurofibromaspheres are engrafted in the sciatic nerve of nude mice. This model constitutes a versatile platform for drug screening and the study of neurofibroma biology. For complete details on the use and execution of this protocol, please refer to Mazuelas et al. (2022).1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-HNK-1/N-CAM (CD57) antibody, Mouse monoclonal, clone VC1.1, purified from hybridoma cell culture