Merck
Class B1 GPCR activation by an intracellular agonist.
Nature (2023)
Kobayashi, et al.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture