Merck
  • 3,5,3'-Triiodo-L-Thyronine Regulates Actin Cytoskeleton Dynamic in The Differentiated PC-12 Cells during Hypoxia through An αvβ3 Integrin.
3,5,3'-Triiodo-L-Thyronine Regulates Actin Cytoskeleton Dynamic in The Differentiated PC-12 Cells during Hypoxia through An αvβ3 Integrin.
Cell journal (2023-05-21)
Tamar Barbakadze, Elisabed Kvergelidze, Judit Bátor, József Szeberényi, David Mikeladze
ABSTRAKT
Thyroid hormones are involved in the pathogenesis of various neurological disorders. Ischemia/hypoxia that induces rigidity of the actin filaments, which initiates neurodegeneration and reduces synaptic plasticity. We hypothesized that thyroid hormones via alpha-v-beta-3 (αvβ3) integrin could regulate the actin filament rearrangement during hypoxia and increase neuronal cell viability. In this experimental study, we analysed the dynamics of actin cytoskeleton according to the G/F actin ratio, cofilin-1/p-cofilin-1 ratio, and p-Fyn/Fyn ratio in differentiated PC-12 cells with/without T3 hormone (3,5,3'-triiodo-L-thyronine) treatment and blocking αvβ3-integrin-antibody under hypoxic conditions using electrophoresis and western blotting methods. We assessed NADPH oxidase activity under the hypoxic condition by the luminometric method and Rac1 activity using the ELISA-based (G-LISA) activation assay kit. The T3 hormone induces the αvβ3 integrin-dependent dephosphorylation of the Fyn kinase (P=0.0010), modulates the G/F actin ratio (P=0.0010) and activates the Rac1/NADPH oxidase/cofilin-1 (P=0.0069, P=0.0010, P=0.0045) pathway. T3 increases PC-12 cell viability (P=0.0050) during hypoxia via αvβ3 integrin-dependent downstream regulation systems. The T3 thyroid hormone may modulate the G/F actin ratio via the Rac1 GTPase/NADPH oxidase/ cofilin1signaling pathway and αvβ3-integrin-dependent suppression of Fyn kinase phosphorylation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
HEPES, ≥99.5% (titration)