Merck
Apoptotic activity of genipin in human oral squamous cell carcinoma in vitro by regulating STAT3 signaling.
Cell biochemistry and function (2023-10-04)
Dong-Guk Park, Bohwan Jin, Won W Lee, Hyun-Ji Kim, Ji-Hoon Kim, Su-Jung Choi, Seong-Doo Hong, Ji-Ae Shin, Sung-Dae Cho
ABSTRAKT
Genipin, a natural compound derived from the fruit of Gardenia jasminoides Ellis, was reported to have activity against various cancer types. In this study, we determined the underlying mechanism for genipin-induced cell death in human oral squamous cell carcinoma (OSCC). The growth-inhibitory effects of genipin in human OSCC cells was examined by the Cell Counting Kit-8 and soft agar assays. The effects of genipin on apoptosis were assessed by nuclear morphological changes by 4',6-diamidino-2-phenylindole staining, measurement of the sub-G1 population, and Annexin V-fluorescein isothiocyanate/propidium iodide double staining. The underlying mechanism of genipin activity was analyzed by western blot analysis, subcellular fractionation of the nucleus and cytoplasm, immunocytochemistry, and quantitative real-time polymerase chain reaction. Genipin inhibited the growth of OSCC cells and induced apoptosis, which was mediated by a caspase-dependent pathway. Genipin reduced the phosphorylation of signal transducer and activator of transcription 3 (STAT3) at Tyr705 and its nuclear localization. Furthermore, inhibition of p-STAT3Tyr705 levels following genipin treatment was required for the reduction of survivin and myeloid cell leukemia-1 (Mcl-1) expression, leading to apoptotic cell death. The genipin-mediated reduction in survivin and Mcl-1 expression was caused by transcriptional and/or posttranslational regulatory mechanisms. The results provide insight into the regulatory mechanism by which genipin induces apoptotic cell death through the abrogation of nuclear STAT3 phosphorylation and suggest that genipin may represent a potential therapeutic option for the treatment of human OSCC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Genipin, ≥98% (HPLC), powder