Merck
International journal of molecular sciences (2023-11-25)
Marine Hanse, Samina Akbar, Hamed Layeghkhavidaki, Frances T Yen
ABSTRAKT
Garcinia cambogia extract (GCE) is a popular weight-loss supplement that also lowers plasma triglyceride (TG) levels. We hypothesized that GCE-mediated inhibition of ATP citrate lyase and thereby hepatic TG production could lead to compensatory mechanisms, including increased hepatic TG uptake via lipoprotein receptors. GCE (20 mg/day) administered 40 days orally to female C57BL/6Rj mice on a standard chow diet led to a decrease in both plasma fasting and post-prandial TG-rich lipoprotein levels, but with no significant change in body weight gain. Lipolysis stimulated lipoprotein receptor (LSR) protein levels, but not those of LDL-receptor, were increased as compared to controls. Mouse Hepa1-6 cells treated with the GCE active ingredient, hydroxycitrate, also led to increased LSR protein levels. Hepatic total cholesterol, TG, and muscle TG contents were higher in GCE-treated animals as compared to controls, whereas adipose TG levels were unchanged. LSR and LDL-receptor protein levels were correlated with liver total cholesterol, but only LDL-receptor was associated with liver TG. These results show that GCE treatment in mice on a standard chow diet led to significantly increased liver and muscle lipids, with no significant change in adipose tissue TG levels, which should be considered in the long-term use of GCE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-LSR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture