Merck
Total Synthesis of (+)-Conolidine by the Gold(I)-Catalyzed Cascade Cyclization of a Conjugated Enyne.
The Journal of organic chemistry (2016-06-09)
Saori Naoe, Yusuke Yoshida, Shinya Oishi, Nobutaka Fujii, Hiroaki Ohno
ABSTRAKT
A total synthesis of (+)-conolidine has been achieved via the gold(I)-catalyzed cascade cyclization of a conjugated enyne. Remarkably, this strategy allowed for the simultaneous formation of the indole ring and the ethylidene-substituted piperidine moiety of (+)-conolidine under homogeneous gold catalysis in an enantioselective manner (88-91% ee).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Conolidine, ≥98% (HPLC)