Merck
  • Dexmedetomidine Stereotaxic Injection Alleviates Neuronal Loss Following Bilateral Common Carotid Artery Occlusion via Up-Regulation of BDNF Expression.
Dexmedetomidine Stereotaxic Injection Alleviates Neuronal Loss Following Bilateral Common Carotid Artery Occlusion via Up-Regulation of BDNF Expression.
In vivo (Athens, Greece) (2023-12-27)
Dongjoon Kim, Yejin Shin, Suyeon Cho, Haksung Kim, Hyoin Hwang, Hyekyoung Shin, Yoonyoung Chung, Yong-Hyun Jun
ABSTRAKT
Neurogenesis is an important process in the recovery from neurological damage caused by ischemic lesions. Endogenous neurogenesis is insufficient to restore neuronal damage following cerebral ischemia. Dexmedetomidine (DEX) exerts neuroprotective effects against cerebral ischemia and ischemia/reperfusion injury. DEX promotes neurogenesis, including neuronal proliferation and maturation in the hippocampus. In a previous study, we showed that early neurogenesis increased 3 days after bilateral common carotid artery occlusion (BCCAO). In this study, we investigated the effect of DEX on neurogenesis 3 days after BCCAO. Male Sprague-Dawley (SD) rats (7-8 weeks old) were used as a BCCAO model. Right and left common carotid arteries of the rats were occluded using 4-0 silk sutures. Two hours after surgery, an intracranial DEX injection was administered to rats that underwent surgery using a stereotaxic injector. Brains were obtained from control and BCCAO rats 3 days after surgery. Immunohistochemistry was performed on the cortex and dentate gyrus of the hippocampus using a NeuN antibody. Western blot was performed with HIF1α and brain-derived neurotrophic factor (BDNF) antibodies. The number of mature neurons decreased 3 days after BCCAO, but DEX treatment alleviated neural loss in the parietal cortex and hippocampus. Up-regulation of BDNF was also observed after dexmedetomidine treatment. Stereotaxic injection of dexmedetomidine alleviates neural loss following BCCAO by up-regulating BDNF expression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dexmedetomidine hydrochloride, ≥98% (HPLC)