Merck
Katanin knockdown supports a role for microtubule severing in release of basal bodies before mitosis in Chlamydomonas.
Molecular biology of the cell (2008-11-14)
M Qasim Rasi, Jeremy D K Parker, Jessica L Feldman, Wallace F Marshall, Lynne M Quarmby
ABSTRAKT
Katanin is a microtubule-severing protein that participates in the regulation of cell cycle progression and in ciliary disassembly, but its precise role is not known for either activity. Our data suggest that in Chlamydomonas, katanin severs doublet microtubules at the proximal end of the flagellar transition zone, allowing disengagement of the basal body from the flagellum before mitosis. Using an RNA interference approach we have discovered that severe knockdown of the p60 subunit of katanin, KAT1, is achieved only in cells that also carry secondary mutations that disrupt ciliogenesis. Importantly, we observed that cells in the process of cell cycle-induced flagellar resorption sever the flagella from the basal bodies before resorption is complete, and we find that this process is defective in KAT1 knockdown cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Supelco
IGEPAL® CA-630