Merck
Structure and activity of human mitochondrial peptide deformylase, a novel cancer target.
Journal of molecular biology (2009-02-25)
Sindy Escobar-Alvarez, Yehuda Goldgur, Guangli Yang, Ouathek Ouerfelli, Yueming Li, David A Scheinberg
ABSTRAKT
Peptide deformylase proteins (PDFs) participate in the N-terminal methionine excision pathway of newly synthesized peptides. We show that the human PDF (HsPDF) can deformylate its putative substrates derived from mitochondrial DNA-encoded proteins. The first structural model of a mammalian PDF (1.7 A), HsPDF, shows a dimer with conserved topology of the catalytic residues and fold as non-mammalian PDFs. The HsPDF C-terminus topology and the presence of a helical loop (H2 and H3), however, shape a characteristic active site entrance. The structure of HsPDF bound to the peptidomimetic inhibitor actinonin (1.7 A) identified the substrate-binding site. A defined S1' pocket, but no S2' or S3' substrate-binding pockets, exists. A conservation of PDF-actinonin interaction across PDFs was observed. Despite the lack of true S2' and S3' binding pockets, confirmed through peptide binding modeling, enzyme kinetics suggest a combined contribution from P2'and P3' positions of a formylated peptide substrate to turnover.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin sodium salt, Ready Made Solution, 100 mg/mL
Sigma-Aldrich
p-Aminobenzamidine–Agarose, (1:1 suspension in a 1.0 M NaCl + 20% ethanol solution)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
p-Aminobenzamidine–Agarose, saline suspension