Merck
A constitutively active form of neurokinin 1 receptor and neurokinin 1 receptor-mediated apoptosis in glioblastomas.
Journal of neurochemistry (2009-06-13)
Toshimasa Akazawa, Shawn G Kwatra, Laura E Goldsmith, Mark D Richardson, Elizabeth A Cox, John H Sampson, Madan M Kwatra
ABSTRAKT
Previous studies have shown that neurokinin 1 receptor (NK1R) occurs naturally in human glioblastomas and its stimulation causes cell proliferation. In the present study we show that stimulation of NK1R in human U373 glioblastoma cells by substance P increases Akt phosphorylation by 2.5-fold, with an EC(50) of 57 nM. Blockade of NK1R lowers basal phosphorylation of Akt, indicating the presence of a constitutively active form of NK1R; similar results are seen in U251 MG and DBTRG-05 glioblastoma cells. Linkage of NK1R to Akt implicates NK1R in apoptosis of glioblastoma cells. Indeed, treatment of serum-starved U373 cells with substance P reduces apoptosis by 53 +/- 1% (p < 0.05), and treatment with NK1R antagonist L-733,060 increases apoptosis by 64 +/- 16% (p < 0.01). Further, the blockade of NK1R in human glioblastoma cells with L-733,060 causes cleavage of Caspase-3 and proteolysis of poly (ADP-ribose) polymerase. Experiments designed to elucidate the mechanism of NK1R-mediated Akt phosphorylation revealed total involvement of non-receptor tyrosine kinase Src and phosphatidyl-3-kinase, a partial involvement of epidermal growth factor receptor, and no involvement of mitogen-activated protein/extracellular signal-related kinase. Taken together, the results of the present study indicate a key role for NK1R in glioblastoma apoptosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).