Merck
  • Identification and expression analysis of CBF/DREB1 and COR15 genes in mutants of Brassica oleracea var. botrytis with enhanced proline production and frost resistance.
Identification and expression analysis of CBF/DREB1 and COR15 genes in mutants of Brassica oleracea var. botrytis with enhanced proline production and frost resistance.
Plant physiology and biochemistry : PPB (2011-10-18)
Fazal Hadi, Martyn Gilpin, Michael P Fuller
ABSTRAKT
Frost resistant mutants of Brassica oleracea var. botrytis were investigated for the presence of CBF/DREB1 and COR15a gene products and induced frost resistance. Total RNA of clones was isolated after 3 h, 6 h, 24 h and 14 d acclimation at 4 °C and proteins and free proline were isolated after 14 d acclimation. cDNA was produced using RT-PCR and the first CBF gene in B. oleracea detected and did quantify. Through SDS-PAGE and Western blotting, the COR15a protein was detected for the first time in B. oleracea. The results confirmed the first report of the presence of BoCBF/DREB1 in B. oleracea and this only appeared under cold acclimation. The sequence analysis of predicted amino acids revealed a very high homology (90%) with CBF sequences of other Brassica species (BnCBF5/DREB1, BrDREB1 and BjDREB1B) and homology reduced to 67% when compared to plants other than Brassicas. BoCBF/DREB1 transcript levels increased up to 24 h acclimation and then declined. Some mutants showed BoCBF/DREB1 expression at 3 h while others only after 6 h and 24 h acclimation. The genotypes showed positive significant correlation between BoCBF/DREB1 expression and frost resistance (R(2) = 0.9343). The proline level under acclimation increased about 8 fold and demonstrated positive and significant correlation with BoCBF/DREB1 expression. Proline also showed positive and significant correlation with frost resistance under cold acclimation but very not under non-acclimation. All clones were positive for COR15a protein after 14 d cold acclimation and expression correlated with frost resistance. Under non-acclimation COR15a was constitutively expressed in 3 mutants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 10 purifications
Sigma-Aldrich
Spectrum Plant Total RNA Kit, sufficient for 50 purifications