Merck
  • Modeling zinc in biomolecules with the self consistent charge-density functional tight binding (SCC-DFTB) method: applications to structural and energetic analysis.
Modeling zinc in biomolecules with the self consistent charge-density functional tight binding (SCC-DFTB) method: applications to structural and energetic analysis.
Journal of computational chemistry (2003-03-13)
Marcus Elstner, Qiang Cui, Petra Munih, Efthimios Kaxiras, Thomas Frauenheim, Martin Karplus
ABSTRAKT
Parameters for the zinc ion have been developed in the self-consistent charge density functional tight-binding (SCC-DFTB) framework. The approach was tested against B3LYP calculations for a range of systems, including small molecules that contain the typical coordination environment of zinc in biological systems (cysteine, histidine, glutamic/aspartic acids, and water) and active site models for a number of enzymes such as alcohol dehydrogenase, carbonic anhydrase, and aminopeptidase. The SCC-DFTB approach reproduces structural and energetic properties rather reliably (e.g., total and relative ligand binding energies and deprotonation energies of ligands and barriers for zinc-assisted proton transfers), as compared with B3LYP/6-311+G** or MP2/6-311+G** calculations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aminopeptidase from Aeromonas proteolytica, lyophilized powder, 50-150 units/mg protein
Sigma-Aldrich
Aminopeptidase His-tagged from Vibrio proteolyticus