Merck
Paenibacillus sp. strain E18 bifunctional xylanase-glucanase with a single catalytic domain.
Applied and environmental microbiology (2010-04-13)
Pengjun Shi, Jian Tian, Tiezheng Yuan, Xin Liu, Huoqing Huang, Yingguo Bai, Peilong Yang, Xiaoyan Chen, Ningfeng Wu, Bin Yao
ABSTRAKT
Xylanases are utilized in a variety of industries for the breakdown of plant materials. Most native and engineered bifunctional/multifunctional xylanases have separate catalytic domains within the same polypeptide chain. Here we report a new bifunctional xylanase (XynBE18) produced by Paenibacillus sp. E18 with xylanase and beta-1,3-1,4-glucanase activities derived from the same active center by substrate competition assays and site-directed mutagenesis of xylanase catalytic Glu residues (E129A and E236A). The gene consists of 981 bp, encodes 327 amino acids, and comprises only one catalytic domain that is highly homologous to the glycoside hydrolase family 10 xylanase catalytic domain. Recombinant XynBE18 purified from Escherichia coli BL21(DE3) showed specificity toward oat spelt xylan and birchwood xylan and beta-1,3-1,4-glucan (barley beta-glucan and lichenin). Homology modeling and molecular dynamic simulation were used to explore structure differences between XynBE18 and the monofunctional xylanase XynE2, which has enzymatic properties similar to those of XynBE18 but does not hydrolyze beta-1,3-1,4-glucan. The cleft containing the active site of XynBE18 is larger than that of XynE2, suggesting that XynBE18 is able to bind larger substrates such as barley beta-glucan and lichenin. Further molecular docking studies revealed that XynBE18 can accommodate xylan and beta-1,3-1,4-glucan, but XynE2 is only accessible to xylan. These results indicate a previously unidentified structure-function relationship for substrate specificities among family 10 xylanases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Glucanase from Aspergillus niger, powder, dark brown, ~1 U/mg
Sigma-Aldrich
β-Glucanase from Trichoderma longibrachiatum