Merck
Polysaccharide-free nucleic acids and proteins of Abelmoschus esculentus for versatile molecular studies.
Molekuliarnaia biologiia (2012-11-02)
A Manoj-Kumar, K N Reddy, M Manjulatha, L Blanco
ABSTRAKT
Abelmoschus esculentus (okra) is one of the polysaccharide rich crop plants. The polysaccharides interfere with nucleic acids and protein isolation thereby affecting the downstream molecular analysis. So, to understand the molecular systematics of okra, high quality DNA, RNA and proteins are essential. In this study we present a method for extracting genomic DNA, RNA and proteins from polysaccharide rich okra tissues. The conventional extraction procedures were integrated with purification treatments with pectinase, RNase and proteinase K, which improved the quality and quantity of DNA as well. Using SDS, additional washes with CIA and NaCl precipitation improved the RNA isolation both quantitatively and qualitatively. Finally, ammonium acetate mediated protein precipitation and re-solubilization increased the quality of total protein extracts from the okra leaves. All of the methods above not only eliminated the impurities but also improved the quality and quantity of nucleic acids and proteins. Further, we subjected these samples to versatile downstream molecular analyses such as restriction endonuclease digestion, RAPD, Southern, reverse transcription-PCR and Western analysis and were proved to be successful.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein