Merck
Simplifying alternating peptide synthesis by protease-catalyzed dipeptide oligomerization.
Chemical communications (Cambridge, England) (2012-11-30)
Xu Qin, Anne C Khuong, Zheng Yu, Wenzhe Du, John Decatur, Richard A Gross
ABSTRAKT
A unique route to perfectly alternating oligopeptides is described. First, AG-OEt was prepared by standard chemical coupling. Then, AG-OEt was converted by papain-catalysis in 30 s to (AG)(x) (80%-yield, x = 9.4 ± 0.3). Extension of this strategy to other alternating peptide compositions as well as the importance of selecting a protease and reaction conditions that avoid competing transamidation and hydrolysis reactions is discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Papain from Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled
Sigma-Aldrich
Papain from papaya latex, crude powder, 1.5-10 units/mg solid