Merck
  • The protective role of yeast cathepsin D in acetic acid-induced apoptosis depends on ANT (Aac2p) but not on the voltage-dependent channel (Por1p).
The protective role of yeast cathepsin D in acetic acid-induced apoptosis depends on ANT (Aac2p) but not on the voltage-dependent channel (Por1p).
FEBS letters (2012-12-12)
Helena Pereira, Flávio Azevedo, António Rego, Maria João Sousa, Susana R Chaves, Manuela Côrte-Real
ABSTRAKT
We have previously shown that the yeast Cathepsin D (CatD) Pep4p translocates from the vacuole to the cytosol during acetic acid-induced apoptosis and is required for efficient mitochondrial degradation, though its specific role in this process is still elusive. Here, we show that the protective role of Pep4p in acetic acid-induced apoptosis depends on its catalytic activity and is independent of the yeast voltage-dependent anion channel Por1p (which has no role on mitochondrial degradation) but dependent on AAC proteins, the yeast adenine nucleotide translocator. Our results demonstrate a differential interplay between yeast vacuolar CatD and mitochondrial proteins involved in apoptosis regulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cathepsin D from bovine spleen, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Cathepsin D from human liver, lyophilized powder, ≥250 units/mg protein (E1%/280)