Merck
  • Phospholipase C of Cryptococcus neoformans regulates homeostasis and virulence by providing inositol trisphosphate as a substrate for Arg1 kinase.
Phospholipase C of Cryptococcus neoformans regulates homeostasis and virulence by providing inositol trisphosphate as a substrate for Arg1 kinase.
Infection and immunity (2013-02-06)
Sophie Lev, Desmarini Desmarini, Cecilia Li, Methee Chayakulkeeree, Ana Traven, Tania C Sorrell, Julianne T Djordjevic
ABSTRAKT
Phospholipase C (PLC) of Cryptococcus neoformans (CnPlc1) is crucial for virulence of this fungal pathogen. To investigate the mechanism of CnPlc1-mediated signaling, we established that phosphatidylinositol 4,5-bisphosphate (PIP(2)) is a major CnPlc1 substrate, which is hydrolyzed to produce inositol trisphosphate (IP(3)). In Saccharomyces cerevisiae, Plc1-derived IP(3) is a substrate for the inositol polyphosphate kinase Arg82, which converts IP(3) to more complex inositol polyphosphates. In this study, we show that in C. neoformans, the enzyme encoded by ARG1 is the major IP(3) kinase, and we further demonstrate that catalytic activity of Arg1 is essential for cellular homeostasis and virulence in the Galleria mellonella infection model. IP(3) content was reduced in the CnΔplc1 mutant and markedly increased in the CnΔarg1 mutant, while PIP(2) was increased in both mutants. The CnΔplc1 and CnΔarg1 mutants shared significant phenotypic similarity, including impaired thermotolerance, compromised cell walls, reduced capsule production and melanization, defective cell separation, and the inability to form mating filaments. In contrast to the S. cerevisiae ARG82 deletion mutant (ScΔarg82) strain, the CnΔarg1 mutant exhibited dramatically enlarged vacuoles indicative of excessive vacuolar fusion. In mammalian cells, PLC-derived IP(3) causes Ca(2+) release and calcineurin activation. Our data show that, unlike mammalian PLCs, CnPlc1 does not contribute significantly to calcineurin activation. Collectively, our findings provide the first evidence that the inositol polyphosphate anabolic pathway is essential for virulence of C. neoformans and further show that production of IP(3) as a precursor for synthesis of more complex inositol polyphosphates is the key biochemical function of CnPlc1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Arginase from bovine liver, Protein ≥70 % by biuret, powder