Merck
Receptor for advanced glycation end products (RAGE) and glyoxalase I gene polymorphisms in pathological pregnancy.
Clinical biochemistry (2012-07-10)
Anna Germanová, Alexandra Muravská, Marie Jáchymová, Zdeněk Hájek, Michal Koucký, Oto Mestek, Tomáš Zima, Marta Kalousová
ABSTRAKT
The aim of the study was to analyze polymorphisms of receptor for advanced glycation end products (RAGE) gene, and glyoxalase I gene and soluble RAGE, sRAGE, in physiological and pathological pregnancy. Polymorphisms of RAGE gene (-429 T/C, -374 T/A, 557 G/A, 2184 A/G) and glyoxalase I gene (A419C) and sRAGE serum levels were determined in 284 women with pathological and physiological pregnancy. No differences in distribution of genotype and allelic frequencies of studied polymorphisms were found. GA genotype of RAGE 557 G/A polymorphism (known as Gly82Ser) is associated with lower sRAGE serum levels in healthy pregnant women compared to GG genotype (483 ± 104 vs. 692 ± 262 pg/mL, p=0.008). sRAGE correlates negatively with ALT in patients with pregnancy intrahepatic cholestasis (r=-0.536, p=0.05). We did not show any association of RAGE and glyoxalase I gene polymorphisms with pathological pregnancy, however further studies are needed to confirm the results.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glyoxalase I from Saccharomyces cerevisiae, Grade IV, buffered aqueous glycerol solution, ≥400 units/mg protein