Merck
  • Proton-in-flight mechanism for the spontaneous hydrolysis of N-methyl O-phenyl sulfamate: implications for the design of steroid sulfatase inhibitors.
Proton-in-flight mechanism for the spontaneous hydrolysis of N-methyl O-phenyl sulfamate: implications for the design of steroid sulfatase inhibitors.
The Journal of organic chemistry (2012-04-11)
David R Edwards, Richard Wolfenden
ABSTRAKT
The hydrolysis of N-methyl O-phenyl sulfamate (1) has been studied as a model for steroid sulfatase inhibitors such as Coumate, 667 Coumate, and EMATE. At neutral pH, simulating physiological conditions, hydrolysis of 1 involves an intramolecular proton transfer from nitrogen to the bridging oxygen atom of the leaving group. Remarkably, this proton transfer is estimated to accelerate the decomposition of 1 by a factor of 10(11). Examination of existing kinetic data reveals that the sulfatase PaAstA catalyzes the hydrolysis of sulfamate esters with catalytic rate accelerations of ~10(4), whereas the catalytic rate acceleration generated by the enzyme for its cognate substrate is on the order of ~10(15). Rate constants for hydrolysis of a wide range of sulfuryl esters, ArOSO(2)X(-), are shown to be correlated by a two-parameter equation based on pK(a)(ArOH) and pK(a)(ArOSO2XH).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis