- Amino acid analysis by HPLC: optimized conditions for chromatography of phenylthiocarbamyl derivatives.
Amino acid analysis by HPLC: optimized conditions for chromatography of phenylthiocarbamyl derivatives.
Analytical biochemistry (1986-05-01)
R F Ebert
PMID3728962
ABSTRAKT
A method for analysis of phenylthiocarbamyl-amino acids by HPLC is described. Solvent A contains 0.05 M sodium acetate plus triethylamine (2.75 ml/liter) adjusted to pH 6.40 with phosphoric acid. Solvent B consists of 50% solvent A, 40% acetonitrile, and 10% methanol. The stationary phase is a 3-micron Spherisorb ODS-2 column (4.6 X 100 mm). A new sample can be injected every 27 min. Variation of the triethylamine concentration from 1 to 4 ml/liter is shown to affect column selectivity in a predictable manner, and this provides a rational basis for optimizing chromatographic conditions. Resolution of certain hydrophilic amino acids (Arg, Thr, Ala, and Pro), and mobile phase stability are improved over previously described procedures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid