Merck
TRPA1 regulates gastrointestinal motility through serotonin release from enterochromaffin cells.
Proceedings of the National Academy of Sciences of the United States of America (2009-02-13)
Katsura Nozawa, Eri Kawabata-Shoda, Hitoshi Doihara, Ryosuke Kojima, Hidetsugu Okada, Shinobu Mochizuki, Yorikata Sano, Kohei Inamura, Hitoshi Matsushime, Tomonobu Koizumi, Toshihide Yokoyama, Hiroyuki Ito
ABSTRAKT
Serotonin (5-hydroxytryptamine; 5-HT) is abundantly present throughout the gastrointestinal tract and stored mostly in enterochromaffin (EC) cells, which are located on the mucosal surface. 5-HT released from EC cells stimulate both intrinsic and extrinsic nerves, which results in various physiological and pathophysiological responses, such as gastrointestinal contractions. EC cells are believed to have the ability to respond to the chemical composition of the luminal contents of the gut; however, the underlying molecular and cellular mechanisms have not been identified. Here, we demonstrate that the transient receptor potential (TRP) cation channel TRPA1, which is activated by pungent compounds or cold temperature, is highly expressed in EC cells. We also found that TRPA1 agonists, including allyl isothiocyanate and cinnamaldehyde, stimulate EC cell functions, such as increasing intracellular Ca(2+) levels and 5-HT release, by using highly concentrated EC cell fractions and a model of EC cell function, the RIN14B cell line. Furthermore, we showed that allyl isothiocyanate promotes the contraction of isolated guinea pig ileum via the 5-HT(3) receptor. Taken together, our results indicate that TRPA1 acts as a sensor molecule for EC cells and may regulate gastrointestinal function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture