Merck
Time- and dose-dependent induction of HSP70 in Lemna minor exposed to different environmental stressors.
Bulletin of environmental contamination and toxicology (2011-06-18)
Stefan Tukaj, Joanna Bisewska, Katarzyna Roeske, Zbigniew Tukaj
ABSTRAKT
The objective of this study was to examine the influence of different stressors, including cadmium (heavy metal), anthracene (polycyclic aromatic hydrocarbon-PAH) and chloridazon (herbicide), on population growth and biosynthesis of cytoplasmic HSP70 in Lemna minor (duckweed) in short (4 h)- and long (7 days)-term tests. A heat shock response was confirmed in Lemna exposed to high temperature: 35, 37.5, 40, or 42.5°C in short-term (4 h) treatments. The chemicals tested stimulated the biosynthesis of the cytoplasmic HSP70 protein in a concentration-dependent way (0.5-5 μM), higher in fronds exposed to lower doses of stressors. Additionally, production of HSP70 was greater after 4 h of incubation than after 7 days. The results suggest that HSP70 could be applied as a non-specific and sensitive detector of stress induced by different chemicals at concentrations below those that produce the type of response observed in classical cytotoxicity tests, such as growth inhibition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diaminobenzidine (DAB) Liquid Substrate System tetrahydrochloride
Sigma-Aldrich
3,3′-Diaminobenzidine (DAB) Liquid Substrate Dropper System, liquid
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Supelco
Anthracene, analytical standard