Merck
  • Simultaneous determination of water and enediols or thiols in chemical products and drugs not amenable to direct Karl Fischer titration.
Simultaneous determination of water and enediols or thiols in chemical products and drugs not amenable to direct Karl Fischer titration.
Journal of AOAC International (1999-09-18)
I Kuselman, F Sherman, T Bourenko, A Shenhar
ABSTRAKT
A method proposed for determining water and enediols or thiols is based on consecutive titration of the enediols or thiols by a novel reagent and of water by the conventional Karl Fischer reagent in the same cell. The time for both titrations is 8-20 min. The novel reagent consists of iodine, sodium acetate as a base, and potassium iodide in a nonaqueous solvent system. The method is applicable for quality control of chemical products and drugs during their production and trade.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
1-Thioglycerol, BioXtra, ≥97%
Sigma-Aldrich
1-Thioglycerol, ≥97%
Sigma-Aldrich
1-Thioglycerol, ≥99.0% (GC)