Merck
  • The measurement of theophylline in human serum or plasma using gas chromatography and isotope dilution-mass spectrometry (GC-IDMS) taking other substituted xanthines into consideration.
The measurement of theophylline in human serum or plasma using gas chromatography and isotope dilution-mass spectrometry (GC-IDMS) taking other substituted xanthines into consideration.
Clinical laboratory (2002-10-23)
Michael Kress, Dieane Meissner, Patricia Kaiser, Rainer Hanke, William Graham Wood
ABSTRAKT
A method is described which uses a combination of gas chromatography and isotope dilution-mass spectrometry (GC-IDMS) to determine the concentration of theophylline (1,3-dimethyl xanthine) in human plasma or serum samples. The effects of similar substituted xanthines - namely theobromine (3,7-dimethyl xanthine), paraxanthine (1,7-dimethyl xanthine) 1,3-dimethyl-7-(2-hydroxyethyl) xanthine (internal standard HPLC) and caffeine (1,3,7-trimethyl xanthine) were tested to confirm the specificity of the method. The derivatisation of all xanthines was performed with N-methyl-N-trimethylsilyl trifluroacetamide (MSTFA). The internal standard used was 2-(13)C ,1,3-(15)N2-theophylline. The extraction and derivatisation procedures were examined in detail and optimised stepwise during the development of the method. High-performance liquid chromatography (HPLC) was used for comparison.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, derivatization grade (GC derivatization), LiChropur, ≥98.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur