Merck
Ionic liquids as advantageous solvents for headspace gas chromatography of compounds with low vapor pressure.
Analytical chemistry (2005-01-15)
M Andre, J Loidl, G Laus, H Schottenberger, G Bentivoglio, K Wurst, K-H Ongania
ABSTRAKT
The potential of ionic liquids as solvents for headspace gas chromatography was investigated. Three compounds with boiling points above 200 degrees C were selected to demonstrate the feasibility of the concept described. 2-Ethylhexanoic acid, formamide, and tri-n-butylamine as examples of acidic, neutral, and basic analytes were dissolved in acidic 1-n-butyl-3-methylimidazolium hydrogen sulfate (1), neutral 1-n-butyl-2,3-dimethylimidazolium dicyanamide (2), and 2 containing 1,8-diazabicyclo[5.4.0]undec-7-ene to adjust basic conditions. All analytes could be determined with limits of detection and limits of quantification in the low-ppm concentration range.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Supelco
2-Ethylhexanoic acid, analytical standard
Sigma-Aldrich
2-Ethylhexanoic acid, 99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
Tributylamine, ≥98.5%
Sigma-Aldrich
Tributylamine, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Tributylamine, puriss. plus, ≥99.5% (GC)
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%