Merck
  • Determination of succinylacetone in dried blood spots and liquid urine as a dansylhydrazone by liquid chromatography tandem mass spectrometry.
Determination of succinylacetone in dried blood spots and liquid urine as a dansylhydrazone by liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-12-31)
Osama Y Al-Dirbashi, Mohamed S Rashed, Herman J Ten Brink, Cornelis Jakobs, Najlaa Filimban, Lujane Y Al-Ahaidib, Minnie Jacob, Moeen M Al-Sayed, Zuhair Al-Hassnan, Eissa Faqeih
ABSTRAKT
Succinylacetone (SA) is a specific marker for the inherited metabolic disease, hepatorenal tyrosinemia. We developed a stable-isotope dilution liquid chromatography tandem mass spectrometry for the determination of SA in dried blood spots (DBS) and liquid urine using a (13)C(4)-SA as internal standard. SA was extracted, converted to the butyl ester and derivatized with dansylhydrazine (Dns-H). Calibration curves in DBS and urine calibrators were linear up to 100 and 30 microM, respectively. At a signal-to-noise ratio of 3, the limits of detection in DBS and urine were 0.2 and 0.005 microM, respectively. Total run time was 5 min. Intra- and inter-assay precision expressed as coefficient of variation were better than 9.1% with more than 96% recovery. The method was applied retrospectively and prospectively for the diagnosis of hepatorenal tyrosinemia and for follow-up of patients under treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dansylhydrazine, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Dansylhydrazine, 98%