Merck
Athlete's foot.
Clinical evidence (2006-04-20)
Fay Crawford
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Naftifine hydrochloride
Sigma-Aldrich
Tioconazole, 97%
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material