Merck
[Determination of three nitroimidazole residues in royal jelly by high performance liquid chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2006-10-05)
Tao Ding, Jinzhong Xu, Chongyu Shen, Yuan Jiang, Huilan Chen, Bin Wu, Zengyun Zhao, Gonghai Li, Jing Zhang, Fei Liu
ABSTRAKT
A method for analysis of trace metronidazole (MTZ), dimetridazole (DMZ) and ronidazole (RNZ) residues in royal jelly was developed by high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). After samples were dissolved in sodium hydroxide solution to disassociate target analytes from matrix, liquid-liquid extraction methods by ethyl acetate solvent were used. Matrix effects were minimized and good quantitation results were obtained by using highly-selective reaction monitoring (H-SRM) technology when deuterated dimetridazole (dimetridazole-D3) was selected as internal standard. Limits of detection (LODs) were 1.0 microg/kg for DMZ, 0.5 microg/kg for MTZ and RNZ (S/N > 5). Limits of quantitation (LOQs) were 2.0 microg/kg for DMZ, 1.0 microg/kg for MTZ and RNZ (S/N > 10). The linear ranges were 2.0 - 200 microg/L for all target analytes. Recoveries and relative standard deviations (RSDs) were in the ranges of 96.6% - 110.6% and 2.1% -7.4%, respectively. This method is suitable for statutory residue testing in the National Residue Surveillance Plan in China and meets the requirement for export.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dimetridazole, VETRANAL®, analytical standard
Supelco
Ronidazole, analytical standard
Supelco
Ipronidazole, VETRANAL®, analytical standard