Merck
Stereoselective synthesis of 2,4,6-trisubstituted tetrahydropyrans by the use of cyclopropanols as homoenols.
Angewandte Chemie (International ed. in English) (2007-03-28)
Hyung Goo Lee, Ivan L Lysenko, Jin Kun Cha
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Trifluoroacetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%