Merck
  • The effect of some curarizing drugs in unanaesthetized man. I. d-Tubocurarine chloride, gallamonium iodide, decamethonium iodide, succinylcholine iodide and its bis-monoethyl-substituted derivative in single or repeated doses. Acta Anaesth. Scandinav. 1957, 1, 15-39.
The effect of some curarizing drugs in unanaesthetized man. I. d-Tubocurarine chloride, gallamonium iodide, decamethonium iodide, succinylcholine iodide and its bis-monoethyl-substituted derivative in single or repeated doses. Acta Anaesth. Scandinav. 1957, 1, 15-39.
Acta anaesthesiologica Scandinavica (2007-08-19)
Henning Poulsen, Willy Hougs
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Decamethonium bromide, Ion-Pair reagent, ≥99%
Sigma-Aldrich
Decamethonium bromide, crystalline