Merck
The partial methylation of methyl beta-D-xylopyranoside with methyl sulfate.
Journal of the American Chemical Society (1949-03-01)
O WINTERSTEINER, A KLINGSBERG
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl β-D-xylopyranoside, ≥99% (GC)