Merck
Auxotrophic-precursor directed biosynthesis of nonribosomal lipopeptides with modified tryptophan residues.
Organic & biomolecular chemistry (2008-03-11)
Bagher Amir-Heidari, Jenny Thirlway, Jason Micklefield
ABSTRAKT
Feeding 5-hydroxy and 5-fluorotryptophan to a Streptomyces coelicolor Trp-auxotrophic strain WH101 results in the production of a number of new calcium-dependent antibiotics (CDAs) possessing modified Trp residues. It is anticipated that this method could be used to modulate the biological properties of Trp-containing nonribosomal peptide natural products, or to generate analogues with useful fluorescent properties for studying biological mechanisms of action.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
5-Fluoro-DL-tryptophan, powder or crystals