Merck
Metabolism and autoradiographic evaluation of [(18)F]FE@CIT: a Comparison with [(123)I]beta-CIT and [(123)I]FP-CIT.
Nuclear medicine and biology (2008-05-17)
Dagmar E Ettlinger, Daniela Häusler, Wolfgang Wadsak, Friedrich Girschele, Karoline M Sindelar, Leonhard-Key Mien, Johanna Ungersböck, Helmut Viernstein, Kurt Kletter, Robert Dudczak, Markus Mitterhauser
ABSTRAKT
Since the late 1980s, cocaine analogues based on the phenyltropane structure, such as [(11)C]CFT and [(123)I]beta-CIT have been used for the imaging of the dopamine transporter. FE@CIT (fluoropropyl ester) and FP-CIT (N-fluoropropyl derivative) are further analogues. The aim of this study was to (1) evaluate and compare the metabolic stability of beta-CIT, FP-CIT and FE@CIT against carboxyl esterases and (2) evaluate selectivity of [(18)F]FE@CIT compared to [(123)I]beta-CIT and [(123)I]FP-CIT using autoradiography. In vitro enzymatic hydrolysis assays were performed using different concentrations of beta-CIT, FE@CIT and FP-CIT with constant concentrations of carboxyl esterase. Autoradiography was performed on coronal 20-microm rat brain sections incubated with different radioactivity concentrations of [(123)I]beta-CIT, [(123)I]FP-CIT or [(18)F]FE@CIT and, additionally, with 3-amino-4-(2-dimethylaminomethyl-phenylsulfanyl)-benzonitrile [serotonin transporter (SERT)] and nisoxetine [norepinephrine transporter (NET)] for blocking experiments. In vitro assays showed Michaelis-Menten constants of 175 micromol (beta-CIT), 183 micromol (FE@CIT) and 521 micromol (FP-CIT). Limiting velocities were 0.1005 micromol/min (beta-CIT), 0.1418 micromol/min (FE@CIT) and 0.1308 micromol/min (FP-CIT). This indicates a significantly increased stability of FP-CIT, whereas carboxyl esterase stability of beta-CIT and FE@CIT showed no significant difference. Autoradiographic analyses revealed a good correlation between dopamine transporter (DAT)-rich regions and the uptake pattern of FE@CIT. Blocking experiments showed a higher DAT selectivity for [(18)F]FE@CIT than for the other two tracers. We found that (1) the metabolic stability of FE@CIT was comparable to that of beta-CIT, whereas FP-CIT showed higher resistance to enzymatic hydrolysis; and (2) the overall uptake pattern of [(18)F]FE@CIT on brain slices was comparable to that of [(123)I]beta-CIT and [(123)I]FPCIT. After blocking of NET and SERT binding, a significantly higher DAT selectivity was observed for [(18)F]FE@CIT. Hence, [(18)F]FE@CIT may be of interest for further clinical application.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nisoxetine hydrochloride, solid, ≥98% (HPLC)