Merck
Cardiac norepinephrine transporter protein expression is inversely correlated to chamber norepinephrine content.
American journal of physiology. Regulatory, integrative and comparative physiology (2008-06-21)
Erica A Wehrwein, Lindsay M Parker, Anna A Wright, John M Spitsbergen, Martin Novotny, Dagmar Babankova, Greg M Swain, Beth A Habecker, David L Kreulen
ABSTRAKT
The cardiac neuronal norepinephrine (NE) transporter (NET) in sympathetic neurons is responsible for uptake of released NE from the neuroeffector junction. The purpose of this study was to assess the chamber distribution of cardiac NET protein measured using [(3)H]nisoxetine binding in rat heart membranes and to correlate NE content to NET amount. In whole mounts of atria, NET was colocalized in nerve fibers with tyrosine hydroxylase (TH) immunoreactivity. NE content expressed as micrograms NE per gram tissue was lowest in the ventricles; however, NET binding was significantly higher in the left ventricle than the right ventricle and atria (P < 0.05), resulting in a significant negative correlation (r(2) = 0.922; P < 0.05) of NET to NE content. The neurotoxin 6-hydroxydopamine, an NET substrate, reduced NE content more in the ventricles than the atria, demonstrating functional significance of high ventricular NET binding. In summary, there is a ventricular predominance of NET binding that corresponds to a high NE reuptake capacity in the ventricles, yet negatively correlates to tissue NE content.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nisoxetine hydrochloride, solid, ≥98% (HPLC)