Merck
A fluorescent probe for zinc ions based on N-methyltetraphenylporphine with high selectivity.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-08-22)
Qiu-Juan Ma, Xiao-Bing Zhang, Yan Zhao, Chun-Yan Li, Zhi-Xiang Han, Guo-Li Shen, Ru-Qin Yu
ABSTRAKT
N-methyl-alpha,beta,gamma,delta-tetraphenylporphine (NMTPPH) has been used to detect trace amount of zinc ions in ethanol-water solution by fluorescence spectroscopy. The fluorescent probe undergoes a fluorescent emission intensity enhancement upon binding to zinc ions in EtOH/H(2)O (1:1, v/v) solution. The fluorescence enhancement of NMTPPH is attributed to the 1:1 complex formation between NMTPPH and Zn(II) which has been utilized as the basis for the selective detection of Zn(II). The linear response range covers a concentration range of Zn(II) from 5.0x10(-7) to 1.0x10(-5)mol/L and the detection limit is 1.5x10(-7)mol/L. The fluorescent probe exhibits high selectivity over other common metal ions except for Cu(II).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%