Merck
Potential role of formaldehyde in the mechanism of action of ascorbigens on the basis of BioArena studies.
Biomedical chromatography : BMC (2008-11-29)
György Kátay, Péter G Ott, Edit Kátay, Donát Magyar, Erno Tyihák
ABSTRAKT
The effect of ascorbigen and 1'-methylascorbigen as a model compound pair was studied on the phytopathogenic bacterium Pseudomonas savastanoi pv. phaseolicola in the BioArena experimental system after overpressured layer chromatography. Results showed a characteristic, strong antibacterial effect of 1'-methylascorbigen and weak effect of ascorbigen present on the adsorbent layer as chromatographic spot. Addition of formaldehyde capture compounds (L-arginine, glutathione, dimedone) partially or totally reduced the antibacterial effect of 1'-methylascorbigen and ascorbigen. On adding Cu(II) ions--which mobilize and coordinate formaldehyde--to the culture medium, the antibacterial effect of both compounds became stronger. It is supposed that the weak antibacterial effect of ascorbigen may have originated from the 1'-methylascorbigen formed in situ on the adsorbent layer by partial enzymatic methylation of ascorbigen.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
5,5-Dimethyl-1,3-cyclohexanedione, derivatization grade (HPLC), for the determination of aldehyde formaldehyde, ≥99.0%
Sigma-Aldrich
5,5-Dimethyl-1,3-cyclohexanedione, 95%