Merck
Dopaminergic modulation of olfactory bulb processing affects odor discrimination learning in rats.
Behavioral neuroscience (2009-07-29)
Olga Escanilla, Courtney Yuhas, David Marzan, Christiane Linster
ABSTRAKT
Olfactory behavioral studies have shown that, when modulated through systemic injections, D1 and D2 receptors have opposing effects on odor discrimination learning. In the present study, cannulated male Sprague-Dawley rats were used to investigate how the modulation of these 2 types of dopaminergic receptors through direct infusion of D1/D2 agonists and antagonists into the olfactory bulb affect olfactory perception. Dopaminergic modulation was locally altered by manipulations of D1 (agonist SKF 82958: 14.6, 43.8, & 143.6 mM; antagonist SCH-23390: 13.4, 40.1, & 60.1 mM) and D2 (agonists quinpirole: 78.2, 117.3, & 156.4 mM; antagonist sulpiride: 0.3, 0.9, & 2.9 mM) receptors during a simultaneous odor discrimination task. The authors found that modulation of D2, but not D1, receptors significantly affected rats' odor discrimination performance. A significant positive correlation between blockade of D2 receptors and discrimination performance, as well as a significant negative correlation between D2 receptor activation and discrimination performance, was observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloro-APB hydrobromide, solid