Merck
Antipain Y, a new antipain analog that inhibits neurotransmitter release from rat dorsal root ganglion neurons.
The Journal of antibiotics (2009-11-17)
Koichi Nakae, Fukiko Kojima, Ryuichi Sawa, Yumiko Kubota, Masayuki Igarashi, Naoko Kinoshita, Hayamitu Adachi, Yoshio Nishimura, Yuzuru Akamatsu
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Antipain, >50000 U/mg