Merck
Polycyclic aromatic hydrocarbon concentration levels on the Korean peninsula between 2006 and 2008.
TheScientificWorldJournal (2010-01-12)
Hang Thi Nguyen, Ki-Hyun Kim, C-J Ma, J-M Oh
ABSTRAKT
Concentrations of seven polycyclic aromatic hydrocarbon (PAH) compounds - benzo(a)anthracene (BaA), chrysene (CHRY), benzo(b)fluoranthene (BbF), benzo(k)fluoranthene (BkF), dibenz(a,h)anthracene (DahA), indeno(1,2,3-cd)pyrene (I123P), and benzo(a)pyrene (BaP) - in air were measured as the sum of gas and particle fractions at 32 monitoring stations dispersed across Korea during a 2-year period (February 2006 to January 2008). The data sets were collected at intervals of 1 day (24 h) per month from each monitoring station. According to our analysis, the spatial distribution of PAH is distinguished by manmade activities between different land use types. Evaluation of total PAH (T-PAH) concentration levels, which were derived by summing up all individual compounds, revealed that the T-PAH value varied on the order of commercial (4.85 + or - 4.40 ng m(-3)) rural (4.42 + or - 2.73 ng m(-3)), industrial (4.27 + or - 1.79 ng m(-3)), greenland (3.09 + or - 3.86 ng m(-3)), and background (2.60 + or - 2.54 ng m(-3)) areas. The PAH values, when compared across seasons, tend to peak consistently during the winter (or spring) due to the active consumption of fossil fuels. The overall results of this study confirm that the pollution status of PAH compounds are clearly discernible not only between areas with different levels of anthropogenic activities, but also between periods with changes in environmental conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Benzo[b]fluoranthene, analytical standard
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard
Dibenz[a,h]anthracene, BCR®, certified reference material
Sigma-Aldrich
Benzo[k]fluoranthene, suitable for fluorescence, ≥99%
Supelco
Benzo[b]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Supelco
Indeno[1,2,3-cd]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibenz[a,h]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Benzo[k]fluoranthene, BCR®, certified reference material
Supelco
Benzo[k]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benz[a]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benz[a]anthracene, analytical standard
Benz[a]anthracene, BCR®, certified reference material
Chrysene, BCR®, certified reference material
Supelco
Chrysene, analytical standard
Sigma-Aldrich
Benz[a]anthracene, 99%
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland