- A facile and efficient synthesis of 3,3-dimethyl isopropylidene proline from (+)-3-carene.
A facile and efficient synthesis of 3,3-dimethyl isopropylidene proline from (+)-3-carene.
The Journal of organic chemistry (2010-01-30)
Latha G Nair, Anil Saksena, Raymond Lovey, Mousumi Sannigrahi, Jesse Wong, Jianshe Kong, Xiaoyong Fu, Viyyoor Girijavallabhan
PMID20108962
ABSTRAKT
A highly efficient and practical route to 3,4-isopropylidene proline I, starting from (+)-3-carene, was developed. The three continuous stereocenters were constructed using the inherent chirality of the starting natural product 2. The overall yield for the 12-step synthesis is 34%. The optimized sequence leading to 1 has been successfully applied on a multigram scale, thereby establishing the practicality of this route.