Merck
Development of a membrane-less dynamic field gradient focusing device for the separation of low-molecular-weight molecules.
Electrophoresis (2010-03-02)
Jeffrey M Burke, Colin D Smith, Cornelius F Ivory
ABSTRAKT
Dynamic field gradient focusing uses an electric field gradient generated by controlling the voltage profile of an electrode array to separate and concentrate charged analytes according to their individual electrophoretic mobilities. This study describes a new instrument in which the electrodes have been placed within the separation channel. The major challenge faced with this device is that when applied voltages to the electrodes are larger than the redox potential of water, electrolysis will occur, producing hydrogen ions (H+) plus oxygen gas on the anodes and hydroxide (OH(-)) plus hydrogen gas on the cathodes. The resulting gas bubbles and pH excursions can cause problems with system performance and reproducibility. An on-column, degassing system that can remove gas bubbles "on-the-fly" is described. In addition, the use of a high capacity, low-conductivity buffer to address the problem of the pH shift that occurs due to the production of H+ on the anodes is illustrated. Finally, the successful separation of three, low-molecular-weight dyes (amaranth, bromophenol blue and methyl red) is described.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %
Supelco
Amaranth, analytical standard
Sigma-Aldrich
Amaranth, Dye content 85-95 %
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Millipore
Methyl Red solution, suitable for microbiology