Merck
[Determination of malachite green and leukomalachite green residues in fish muscle by high-performance liquid chromatography method].
Roczniki Panstwowego Zakladu Higieny (2009-01-01)
Jelena Doroszkiewicz-Fiedoruk, Jolanta Masłowiecka, Lech Rodziewicz
ABSTRAKT
Malachite green (MG) and leukomalachite green (LMG) are subjected to monitoring fish muscle, with a minimum required performance limit (MRPL) set 2 microg/kg. Samples were extracted with acetonitryle-buffer mixture and cleaned up on SCX solid phase extraction (SPE) column. LC separation MG and LMG was done on column Luna Phenyl-Hexyl Phenomenex. Samples were fortified with MG and LMG between 2 - 25 microg/kg. The coefficients of variation (CV%) were lower than 14% for MG and 16%for LMG. The mean recoveries were in the range 65- 83% for MG and 70- 73% for LMG. This method fulfils the criteria for identification and determination of MG andLMG residues in the samples of fish muscle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Leucomalachite Green, analytical standard
Sigma-Aldrich
Leucomalachite Green, powder